Learn More
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1 trimethylchlorosilane, ≥97%, MilliporeSigma™ Supelco™
Silylation can be performed by adding N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MTFA) during analysis of fluorometabolites via gas chromatography-mass spectrometry (GC-MS).
Supplier:  Merck Emd Millipore 694785MLF
 View more versions of this product
Catalog No. 111018572
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
24589-78-4
26°C - closed cup
MFCD00000411
5 mL
≥97%
LiChropur
C6H12F3 NOSi
Organic Standard
199.25
GC Derivatization
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany