missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-Methyl-bis(trifluoroacetamide), 99.0%, MilliporeSigma™ Supelco™

Supplier:  Merck Emd Millipore 6594325ML

 View more versions of this product

Catalog No. 111017993


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

685-27-8
1.547 g/mL (at 25°C)
CF3 CON(CH3)COCF3
121°C to 122°C
Organic Standard
223.07
GC/MS (SIM)
LiChropur
45°C
MFCD00000412
MBTFA
25 mL
≥99% (GC)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany