missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Methyl-bis(trifluoroacetamide), 99.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 685-27-8 |
| Product Line | LiChropur |
| Density | 1.547 g/mL (at 25°C) |
| Flash Point | 45°C |
| Linear Formula | CF3 CON(CH3)COCF3 |
| MDL Number | MFCD00000412 |
| Boiling Point | 121°C to 122°C |
| Synonym | MBTFA |
| Type | Organic Standard |
| Quantity | 5 mL |
| Show More |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany