accessibility menu, dialog, popup

UserName

H3CNOFFFOFFF

CAS RN 685-27-8

IUPAC Name: 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
Synonyms: 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
Molecular Weight (g/mol): 223.07
Molecular Formula: C5H3F6NO2
InChi Key: AWGBWLXGUPTXHF-UHFFFAOYSA-N
SMILES: CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)