missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Isobutyryl-L-cysteine, For Chiral Derivatization, 97.0%, MilliporeSigma™ Supelco™
Chiral derivatizing agent employed in the assay of the enantiomeric purity of amino acids with OPA
$224.03 - $723.11
Chemical Identifiers
| CAS | 124529-02-8 |
|---|---|
| Molecular Formula | C7H13NO3S |
| Molecular Weight (g/mol) | 191.25 |
| MDL Number | MFCD00155635 |
| InChI Key | BWBQXMAXLAHHTK-YFKPBYRVSA-N |
| Synonym | N-(2-Methylpropionyl)-L-cysteine |
| IUPAC Name | (2R)-2-(2-methylpropanamido)-3-sulfanylpropanoic acid |
| SMILES | CC(C)C(=O)N[C@@H](CS)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
111012831
|
Merck Emd Millipore
58698250MGF |
250 mg | Bottomless Glass Bottle with Fused Cone |
Each for $224.03
|
|
||||
|
111012830
|
Merck Emd Millipore
586981GF |
1 g | Bottomless Glass Bottle with Fused Cone |
Each for $723.11
|
|
||||
Chemical Identifiers
| 124529-02-8 | |
| 191.25 | |
| BWBQXMAXLAHHTK-YFKPBYRVSA-N | |
| (2R)-2-(2-methylpropanamido)-3-sulfanylpropanoic acid |
| C7H13NO3S | |
| MFCD00155635 | |
| N-(2-Methylpropionyl)-L-cysteine | |
| CC(C)C(=O)N[C@@H](CS)C(O)=O |
Specifications
| 124529-02-8 | |
| Bottomless Glass Bottle with Fused Cone | |
| C7H13NO3S | |
| 250 mg | |
| BWBQXMAXLAHHTK-YFKPBYRVSA-N | |
| (2R)-2-(2-methylpropanamido)-3-sulfanylpropanoic acid | |
| 191.25 | |
| For chiral derivatization |
| 97°C to 101°C | |
| C7H13NO3S | |
| MFCD00155635 | |
| N-(2-Methylpropionyl)-L-cysteine | |
| CC(C)C(=O)N[C@@H](CS)C(O)=O | |
| 191.25 | |
| ≥97.0% |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany