accessibility menu, dialog, popup

UserName

CAS RN 124529-02-8

H3CH3COHN(R)SHHOO

CAS RN 124529-02-8

IUPAC Name: (2R)-2-(2-methylpropanamido)-3-sulfanylpropanoic acid
Synonyms: (2R)-2-(2-methylpropanamido)-3-sulfanylpropanoic acid
Molecular Weight (g/mol): 191.25
Molecular Formula: C7H13NO3S
InChi Key: BWBQXMAXLAHHTK-YFKPBYRVSA-N
SMILES: CC(C)C(=O)N[C@@H](CS)C(O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)