missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-Isobutyryl-L-cysteine, For Chiral Derivatization, 97.0%, MilliporeSigma™ Supelco™

Catalog No. 111012831
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Bottomless Glass Bottle with Fused Cone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111012831 250 mg Bottomless Glass Bottle with Fused Cone
111012830 1 g Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012831 Supplier Merck Emd Millipore Supplier No. 58698250MGF

Chiral derivatizing agent employed in the assay of the enantiomeric purity of amino acids with OPA

Chemical Identifiers

CAS 124529-02-8
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00155635
InChI Key BWBQXMAXLAHHTK-YFKPBYRVSA-N
Synonym N-(2-Methylpropionyl)-L-cysteine
IUPAC Name (2R)-2-(2-methylpropanamido)-3-sulfanylpropanoic acid
SMILES CC(C)C(=O)N[C@@H](CS)C(O)=O

Specifications

Melting Point 97°C to 101°C
Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C7H13NO3S
Quantity 250 mg
Formula Weight 191.25
Percent Purity ≥97.0%
Grade For chiral derivatization

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany