missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ 2,2,3,3,3-Pentafluoro-1-propanol, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014175
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014175 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014175 Supplier Merck Emd Millipore Supplier No. 7987910ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00004673
Synonym PFPOH

Specifications

CAS 422-05-9
Density 1.505 g/mL (at 25°C)
Boiling Point 80°C (748 mmHg)
Linear Formula CF3 CF2 CH2 OH
Quantity 10 mL
Formula Weight 150.05
Percent Purity ≥98.5% (GC)
Grade For GC derivatization
Product Line LiChropur
RTECSNumber UB8800000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany