missing translation for 'onlineSavingsMsg'
Learn More

Lewatit™ MonoPlus TP 214 Ion Exchange Resin, MilliporeSigma™ Supelco™

Catalog No. 111012445
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012445 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012445 Supplier Merck Emd Millipore Supplier No. 62107100G

Lewatit™ MonoPlus TP 214 is a type of chelating ion-exchange resin.

Specifications

CAS 109945-55-3
Product Line Lewatit
Flash Point >250°C
Physical Form Macroporous
MDL Number MFCD01867339
Type Organic Standard
Quantity 100 g

Lewatit is a registered trademark of Lanxess Deutschland GmbH