missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Levocetirizine dihydrochloride

Catalog No. AC467470010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467470010 1 g
AC467470050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467470010 Supplier Thermo Scientific Chemicals Supplier No. 467470010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130018-87-0
Molecular Formula C21H25ClN2O3·2HCl

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble.
Melting Point 214.0°C
Formula Weight 461.81
InChI Key PGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILES Cl.Cl.OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
IUPAC Name 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dihydrochloride
Molecular Weight (g/mol) 461.81
Chemical Name or Material Levocetirizine dihydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Very toxic to aquatic life.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only