missing translation for 'onlineSavingsMsg'
Learn More

Leucovorin Calcium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013178
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013178 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013178 Supplier Merck Emd Millipore Supplier No. PHR15411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

Packaging Ampule
CAS 1492-18-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H21 CaN7O7
Synonym Folinic acid calcium salt; Calcium folinate
Type Certified Reference Material
Quantity 1 g
Formula Weight 511.50
Grade Certified Reference Material