missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Letrozole

Catalog No. 438250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
438250 50 mg
438210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 438250 Supplier Tocris Bioscience™ Supplier No. 4382/50

Potent, reversible non-steroidal aromatase inhibitor

Potent, reversible non-steroidal aromatase inhibitor (IC50 = 11.5 nM). Displays antitumor effects in several animal models. Suppresses the endogenous aromatase-induced proliferation of MCF-7 cells.

Chemical Identifiers

CAS 112809-51-5
Molecular Formula C17H11N5

Specifications

Quantity 50 mg
Formula Weight 285.3
Purity >99%
Molecular Weight (g/mol) 285.31
Chemical Name or Material Letrozole
Recommended Storage Store at +4°C