missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Letrozole
Description
Potent, reversible non-steroidal aromatase inhibitor (IC50 = 11.5 nM). Displays antitumor effects in several animal models. Suppresses the endogenous aromatase-induced proliferation of MCF-7 cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 285.3 |
| Purity | >99% |
| Molecular Weight (g/mol) | 285.31 |
| Chemical Name or Material | Letrozole |
Safety and Handling
| Recommended Storage | Store at +4°C |