missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lercanidipine Hydrochloride 98.0+%, TCI America™
$29.83 - $82.02
Chemical Identifiers
| CAS | 132866-11-6 |
|---|---|
| Molecular Formula | C36H42ClN3O6 |
| Molecular Weight (g/mol) | 648.197 |
| MDL Number | MFCD07773089 |
| InChI Key | WMFYOYKPJLRMJI-UHFFFAOYSA-N |
| Synonym | lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen |
| PubChem CID | 157917 |
| IUPAC Name | 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride |
| SMILES | CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L0282200MG
|
TCI America
L0282200MG |
200 mg |
Each for $29.83
|
|
|||||
|
L02821G
|
TCI America
L02821G |
1 g |
Each for $82.02
|
|
|||||
Chemical Identifiers
| 132866-11-6 | |
| 648.197 | |
| WMFYOYKPJLRMJI-UHFFFAOYSA-N | |
| 157917 | |
| CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl |
| C36H42ClN3O6 | |
| MFCD07773089 | |
| lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen | |
| 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride |
Specifications
| 132866-11-6 | |
| Yellow | |
| MFCD07773089 | |
| lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen | |
| CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl | |
| 648.197 | |
| 648.20 | |
| Crystalline Powder |
| 195°C | |
| C36H42ClN3O6 | |
| 200 mg | |
| WMFYOYKPJLRMJI-UHFFFAOYSA-N | |
| 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride | |
| 157917 | |
| ≥98.0% (HPLC,T) | |
| Lercanidipine Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator