missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lercanidipine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America L02821G
Specifications
| Lercanidipine Hydrochloride | |
| 195°C | |
| C36H42ClN3O6 | |
| 1 g | |
| WMFYOYKPJLRMJI-UHFFFAOYSA-N | |
| 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride | |
| 157917 | |
| ≥98.0% (HPLC,T) |
| 132866-11-6 | |
| Yellow | |
| MFCD07773089 | |
| lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen | |
| CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl | |
| 648.197 | |
| 648.20 | |
| Crystalline Powder |
Chemical Identifiers
| 132866-11-6 | |
| 648.197 | |
| WMFYOYKPJLRMJI-UHFFFAOYSA-N | |
| 157917 | |
| CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl |
| C36H42ClN3O6 | |
| MFCD07773089 | |
| lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen | |
| 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator