missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Lentil Lectin Sepharose™ 4B Medium

Catalog No. 45000070
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000070 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000070 Supplier Cytiva Supplier No. 17044401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Routinely used in the preparation and purification of a number of glycoproteins and carbohydrate-containing molecules

  • Lentil lectin coupled to Sepharose 4B via cyanogen bromide activation
  • Lentil lectin is a metalloprotein, containing Ca2+ and Mn2+, which binds reversibly to polysaccharides and glycoconjugates containing glucose or mannose type sugars
  • Retains binding in 1% deoxycholate; especially suited for purification of detergent-solubilized membrane proteins
  • Ligand concentration 2 mg Lentil lectin/ml; binding capacity for porcine thyroglobulin approx. 16mg/mL medium
  • Average particle size 90μm
  • Stable in the pH range 3-10, and in the presence of all commonly used buffers (Chelating agents and 8M urea to be avoided)
  • Has more equal affinity for α-D-mannose and α-D-glucose compared to Con A

Order Info

Supplied in 20% ethanol containing 0.9% NaCl, 1mM Ca2+ and 1mM Mn2+

Specifications

Binding Capacity 25mg/mL
Ligand Concentration 2mg Lentil lectin/mL Medium
Matrix 4% Agarose
Particle Size 90 μm
pH 3 to 10
Quantity 25 mL
Content And Storage 2°C to 8°C
For Use With (Application) Used for purification of a number of Glycoproteins and other Carbohydrate containing molecules
Ligand Lentil lectin