missing translation for 'onlineSavingsMsg'
Learn More

UVP LED White Light Plate for UVP Imagers

Numéro de catalogue. uvp95047601
Change view
Click to view available options
For Use With (Equipment):
UVP Imagers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. For Use With (Equipment)
UVP95047601 UVP Imagers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. UVP95047601 Fournisseur UVP Code fournisseur 95047601
Il en reste null

For colorimetric imaging in the darkroom via uniform, backlit white light

  • View autoradiographs, as well as Coomassie Blue and Silver stains
  • Dimensions: 11W x 11D in. (28 x 28cm)
  • For use with UVP™ Imagers

Spécifications

For Use With (Equipment) UVP Imagers
For Use With (Application) For colorimetric imaging in the darkroom via uniform, backlit white light
Color White
Product Type Uv Light Plate