missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lecithin, MP Biomedicals™
Description
Molecular biology, Cell biology
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 100 g |
| Formula Weight | 327.27 |
| Physical Form | Solid |
| Chemical Name or Material | Lecithin |
Sécurité et manipulation
| Recommended Storage | Store at 0°C. |