missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MP Biomedicals™

Catalog No. MP215153990
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215153990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215153990 Supplier MP Biomedicals Inc Supplier No. 0215153990

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as an emulsifier and solubilizer for drugs in aqueous solution and as a vehicle for liposomes. Lecithin, MP Biomedicals is a major structural phospholipid in the brain, comprising ∼15% of total lipid and localized to gray matter. It is a purified

Lecithin (aka phosphatidylcholine) is the major structural component of cellular membranes and serves as a reservoir for several lipid messengers. It is the source of the bioactive lipids: lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, platelet activating factor and arachidonic acid.

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Quantity 500 g
Percent Purity ≥90%
Grade Water Dispersible
Physical Form Solid
Recommended Storage Store at -20°C.