missing translation for 'onlineSavingsMsg'
Learn More

Lead Stock Solution, 100 ppm Pb, for USP Heavy Metals Testing, Ricca Chemical

Catalog No. 429616
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
429616 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 429616 Supplier Ricca Chemical Company Supplier No. 429616

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

USP Heavy Metals Testing

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 100ppm Pb
CAS Min % 97.0, 0.46, 0.17, 0.01
CAS Max % 100.0, 0.47, 0.17, 0.01
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Lead Stock Solution
Show More Show Less