missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Phthalocyanine (purified by sublimation) 98.0+%, TCI America™

Numéro de catalogue. L0230200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
L0230200MG 200 mg
L02301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. L0230200MG Fournisseur TCI America Code fournisseur L0230200MG
Il en reste null

Identifiants chimiques

CAS 15187-16-3
Molecular Formula C32H16N8Pb
MDL Number MFCD00053951
Synonym Phthalocyanine Lead(II)

Spécifications

Color Blue-Purple
Quantity 200 mg
UN Number 2291
Formula Weight 719.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lead(II) Phthalocyanine (purified by sublimation)