missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Chloride (purified by sublimation) [for Perovskite precursor], TCI America™

Catalog No. L02911G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L02911G Supplier TCI America Supplier No. L02911G

Chemical Identifiers

CAS 7758-95-4
Molecular Formula Cl2Pb
Molecular Weight (g/mol) 278.10
MDL Number MFCD00011157
InChI Key HWSZZLVAJGOAAY-UHFFFAOYSA-L
Synonym lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
PubChem CID 24459
IUPAC Name λ²-lead(2+) dichloride
SMILES [Cl-].[Cl-].[Pb++]

Specifications

Melting Point 501°C
Color White
Boiling Point 951°C
Quantity 1 g
UN Number 2291
Formula Weight 278.10
Physical Form Crystalline Powder
Chemical Name or Material Lead(II) Chloride (purified by sublimation) [for Perovskite precursor]
EINECSNumber (1)-0252
RTECSNumber OF9450000
TSCA Yes