Learn More
Lead Acetate, For Arsenic, 10% (w/v), LabChem™
$161.75 - $246.43
Chemical Identifiers
| CAS | 6080-56-4 |
|---|---|
| Molecular Formula | C4H12O7Pb |
| Molecular Weight (g/mol) | 379.30 |
| MDL Number | MFCD00150023 |
| InChI Key | MCEUZMYFCCOOQO-UHFFFAOYSA-L |
| Synonym | lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate |
| PubChem CID | 22456 |
| ChEBI | CHEBI:33112 |
| IUPAC Name | λ²-lead(2+) diacetate trihydrate |
| SMILES | O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Chemical Identifiers
| 6080-56-4 | |
| 379.30 | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| 22456 | |
| λ²-lead(2+) diacetate trihydrate |
| C4H12O7Pb | |
| MFCD00150023 | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| CHEBI:33112 | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Specifications
| Colorless | |
| 90,10 | |
| Pb(COOCH3)2·3H2O | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| λ²-lead(2+) diacetate trihydrate | |
| 22456 | |
| 379.33 | |
| Passes Test | |
| Lead oxide; Carbon monoxide; Carbon dioxide | |
| Liquid |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
May cause damage to organs (kidneys, liver, blood, brain) through prolonged or repeated exposure (oral).
Toxic to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Get medical advice and attention if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature