missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate, For Arsenic, 10% (w/v), LabChem™

Catalog No. LC161002
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC161002 1 L
LC161001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC161002 Supplier LabChem Supplier No. LC161002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6080-56-4 , 7732-18-5
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ²-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless
CAS Max % 90, 10
Linear Formula Pb(COOCH3)2·3H2O
Solubility Information Soluble in water
Formula Weight 379.33
Percent Purity 10% (w/v)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Lead oxide; Carbon monoxide; Carbon dioxide
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Lead Acetate, For Arsenic
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
May cause damage to organs (kidneys, liver, blood, brain) through prolonged or repeated exposure (oral).
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Get medical advice and attention if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature