missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Acetate, 2% (w/v), Ricca Chemical
$232.63 - $232.63
Chemical Identifiers
| CAS | 6080-56-4 |
|---|---|
| Molecular Formula | C4H12O7Pb |
| Molecular Weight (g/mol) | 379.30 |
| MDL Number | MFCD00150023 |
| InChI Key | MCEUZMYFCCOOQO-UHFFFAOYSA-L |
| Synonym | lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate |
| PubChem CID | 22456 |
| IUPAC Name | λ2-lead(2+) diacetate trihydrate |
| SMILES | O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Chemical Identifiers
| 6080-56-4 | |
| 379.30 | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| 22456 | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
| C4H12O7Pb | |
| MFCD00150023 | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| λ2-lead(2+) diacetate trihydrate |
Specifications
| Colorless | |
| 1.92,96.0,0.1 | |
| C4H12O7Pb | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O | |
| 379.30 | |
| 2% | |
| Natural Poly Bottle | |
| 1 L | |
| 2% (w/v) |
| 6080-56-4,7732-18-5,64-19-7 | |
| 2.0,100.0,0.1 | |
| MFCD00150023 | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| λ2-lead(2+) diacetate trihydrate | |
| 22456 | |
| Laboratory | |
| Aqueous Solution | |
| Liquid | |
| Lead Acetate |