Learn More
Lead AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Pb), LabChem™
Supplier: LabChem LC161301
Specifications
| Lead AA Standard | |
| Poly Bottle | |
| 97.82,2.02,0.16 | |
| 1g/mL | |
| N2O6Pb | |
| Pb(NO3)2 | |
| UN3264 | |
| Soluble in water | |
| [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 331.20 | |
| 24924 | |
| 331.21 |
| 1000ppm (1mL = 1mg Pb) | |
| 10099-74-8 , 7697-37-2 , 10099-74-8 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Pb) | |
| MFCD00011153 | |
| Colorless | |
| RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
| λ²-lead(2+) dinitrate | |
| 500 mL | |
| CHEBI:37187 | |
| Certified |
Chemical Identifiers
| 10099-74-8 | |
| 331.20 | |
| RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
| CHEBI:37187 | |
| [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| N2O6Pb | |
| MFCD00011153 | |
| 24924 | |
| λ²-lead(2+) dinitrate |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature