missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ LDPE Carboy

Numéro de catalogue. 14388174
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (Metric)
14388174 10 L
14388175 20 L
14388176 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 14388174 Fournisseur Fisherbrand™ Code fournisseur 14388174
Il en reste null

Store and transport reagents; Closure Material: Polypropylene; Color: White

  • Store and transport reagents with 10 L FISHERBRAND LDPE, Carboy, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands.
  • These carboys accept 13.5 rubber stoppers for an alternative sealing method.
  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof
  • Includes:White polypropylene screw cap and 83B closure

Spécifications

Autoclavable Not Autoclavable
Capacity (Metric) 10 L
Color Natural
Disposable Reusable
Material LDPE
Mouth Style Wide
Product Type Carboy
Sterility Non-sterile