missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCN15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150177
Description
LCN15 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The LCN15 (Lipocalin 15) gene encodes a protein that belongs to the lipocalin family, a group of small extracellular proteins characterized by their ability to bind and transport small hydrophobic molecules. LCN15 is a human-specific protein found abundantly in the olfactory mucus. It has been identified as a biomarker for Bowman gland cells in the olfactory neuroepithelium, with its expression correlating with non-degenerated areas of this tissue. The lipocalin family proteins, including LCN15, exhibit significant sequence heterogeneity but possess highly conserved tertiary structures with an eight-stranded antiparallel β-barrel. This structure forms a binding pocket that allows them to transport a wide variety of molecules, including retinol, steroids, and various lipids.Identifiants chimiques
| LCN15 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6UWW0 | |
| 389812 | |
| Recombinant Lipocalin 15 (LCN15) (Pro26-Leu170). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| LCN15 | |
| LCN15; lipocalin 15; lipocalin-15; MSFL2541; PRO6093; UNQ2541; UNQ2541/PRO6093 | |
| LCN15 | |
| Primary | |
| Protein A |
Spécifications
| LCN15 | |
| Polyclonal | |
| Unconjugated | |
| LCN15 | |
| LCN15; lipocalin 15; lipocalin-15; MSFL2541; PRO6093; UNQ2541; UNQ2541/PRO6093 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 389812 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6UWW0 | |
| LCN15 | |
| Recombinant Lipocalin 15 (LCN15) (Pro26-Leu170). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |