Learn More
Lazar Research Laboratories Micro Combination pH Electrode
Catalog No. 136446
Change view
Click to view available options
Connector Types:
BNC
Catalog No. Connector Types
136446 BNC
1 options
Catalog No. 136446 Supplier Lazar Research Laboratories Supplier No. PHR 146B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Comes complete with filling solution and instructions.
Unbreakable combination electrode is ideal for measuring samples as small as 10μL. Tip diameter is under 1mm, for convenient use in microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes. Has a pH range of 1 to 10, temperature range of 0° to 75°C (32° to 167°F), immersion depth of 0.5mm, so you can use it on such flat surfaces as filter paper and agar gels.
Order Info
Available with BNC or U.S. Standard and pin connectors.
Specifications
Connector Types BNC
pH Range 1 to 10