missing translation for 'onlineSavingsMsg'
Learn More

Lauryl chloride, 99%

Catalog No. AC257102500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257102500 250 mL Glass bottle
AC257100010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC257102500 Supplier Thermo Scientific Chemicals Supplier No. 257102500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-52-7 | C12H25Cl | 204.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-52-7
Molecular Formula C12H25Cl
Molecular Weight (g/mol) 204.78
MDL Number MFCD00013688
InChI Key YAYNEUUHHLGGAH-UHFFFAOYSA-N
Synonym lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk
PubChem CID 8192
IUPAC Name 1-chlorododecane
SMILES CCCCCCCCCCCCCl

Specifications

Melting Point -9.3°C
Color Colorless
Density 0.8700g/mL
Boiling Point 260°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4420 to 1.444
Linear Formula CH3(CH2)11Cl
Quantity 250 mL
Specific Gravity 0.87
Solubility Information Solubility in water: 0.0068g/L (20°C). Other solubilities: miscible with most common organic solvents,slightly miscible with alcohol,soluble in ethanol,benzene,ether,miscible with acetone
Formula Weight 204.79
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Lauryl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 203-981-5
TSCA TSCA

RUO – Research Use Only