missing translation for 'onlineSavingsMsg'
Learn More

Lauronitrile 98.0+%, TCI America™

Catalog No. p-7131824
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L011025ML 25 mL
L0110500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L011025ML Supplier TCI America Supplier No. L011025ML

Chemical Identifiers

CAS 2437-25-4
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.32
MDL Number MFCD00001946
InChI Key VXCUURYYWGCLIH-UHFFFAOYSA-N
Synonym lauronitrile, undecyl cyanide, dodecanonitrile, 1-cyanoundecane, n-undecyl cyanide, lauric nitrile, decylacetonitrile, lauric acid nitrile, dodecane nitrile, nitrile 12
PubChem CID 17092
IUPAC Name dodecanenitrile
SMILES CCCCCCCCCCCC#N

Specifications

Color Yellow
Boiling Point 276°C
Quantity 25 mL
Formula Weight 181.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Lauronitrile
EINECSNumber (2)-1507
RTECSNumber JR2600000
TSCA Yes