missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laurocapram 96.0+%, TCI America™
$82.02 - $275.87
Chemical Identifiers
| CAS | 59227-89-3 |
|---|---|
| Molecular Formula | C18H35NO |
| Molecular Weight (g/mol) | 281.484 |
| MDL Number | MFCD00190580 |
| InChI Key | AXTGDCSMTYGJND-UHFFFAOYSA-N |
| Synonym | 1-Dodecylazepan-2-one, N-Dodecyl-epsilon-caprolactam |
| PubChem CID | 42981 |
| IUPAC Name | 1-dodecylazepan-2-one |
| SMILES | CCCCCCCCCCCCN1CCCCCC1=O |
Chemical Identifiers
| 59227-89-3 | |
| 281.484 | |
| AXTGDCSMTYGJND-UHFFFAOYSA-N | |
| 42981 | |
| CCCCCCCCCCCCN1CCCCCC1=O |
| C18H35NO | |
| MFCD00190580 | |
| 1-Dodecylazepan-2-one, N-Dodecyl-epsilon-caprolactam | |
| 1-dodecylazepan-2-one |
Specifications
| 59227-89-3 | |
| Yellow | |
| C18H35NO | |
| 5 mL | |
| AXTGDCSMTYGJND-UHFFFAOYSA-N | |
| 1-dodecylazepan-2-one | |
| 42981 | |
| ≥96.0% (GC) | |
| Laurocapram |
| -7°C | |
| 160°C | |
| MFCD00190580 | |
| 1-Dodecylazepan-2-one, N-Dodecyl-epsilon-caprolactam | |
| CCCCCCCCCCCCN1CCCCCC1=O | |
| 281.484 | |
| 281.48 | |
| Liquid |
Safety and Handling
RTECSNumber : CM3995000
TSCA : No