missing translation for 'onlineSavingsMsg'
Learn More

Laurocapram 96.0+%, TCI America™

Supplier:  TCI America L031725ML

 View more versions of this product

Catalog No. L031725ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Laurocapram
-7°C
160°C
MFCD00190580
1-Dodecylazepan-2-one, N-Dodecyl-epsilon-caprolactam
CCCCCCCCCCCCN1CCCCCC1=O
281.484
281.48
Liquid
59227-89-3
Yellow
C18H35NO
25 mL
AXTGDCSMTYGJND-UHFFFAOYSA-N
1-dodecylazepan-2-one
42981
≥96.0% (GC)

Chemical Identifiers

59227-89-3
281.484
AXTGDCSMTYGJND-UHFFFAOYSA-N
42981
CCCCCCCCCCCCN1CCCCCC1=O
C18H35NO
MFCD00190580
1-Dodecylazepan-2-one, N-Dodecyl-epsilon-caprolactam
1-dodecylazepan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CM3995000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions