missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauric Anhydride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 42°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 382.63 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lauric Anhydride |
Safety and Handling
| TSCA | No |