missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauric Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 143-07-7 |
| Linear Formula | CH3(CH2)10 COOH |
| MDL Number | MFCD00002736 |
| Synonym | Dodecanoic acid; Lauric acid; NSC 5026 |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 200.32 |
| Grade | Certified Reference Material |