missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lauric acid, 99%

Catalog No. AC167281000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167281000 100 g Plastic bottle
AC167280050 5 g Glass bottle
AC167285000 500 g Plastic bottle
AC167280010 1 kg Plastic bottle
AC167280051 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC167281000 Supplier Thermo Scientific Chemicals Supplier No. 167281000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143-07-7 | C12H24O2 | 200.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143-07-7
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00002736
InChI Key POULHZVOKOAJMA-UHFFFAOYSA-N
Synonym lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
PubChem CID 3893
ChEBI CHEBI:30805
IUPAC Name dodecanoic acid
SMILES CCCCCCCCCCCC(=O)O

Specifications

Melting Point 44.0°C to 46.0°C
Color White
Boiling Point 225.0°C (100.0 mmHg)
Flash Point 156°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)10CO2H
Quantity 100 g
Beilstein 02, 359
Merck Index 15, 5439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,ether and alcohol
Formula Weight 200.32
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Lauric acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 205-582-1

RUO – Research Use Only