missing translation for 'onlineSavingsMsg'
Learn More

Lauramide 96.0+%, TCI America™

Catalog No. L007725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L007725G 25 g
L0077100G 100 g
L0077500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L007725G Supplier TCI America Supplier No. L007725G

Chemical Identifiers

CAS 1120-16-7
Molecular Formula C12H25NO
Molecular Weight (g/mol) 199.338
MDL Number MFCD00025532
InChI Key ILRSCQWREDREME-UHFFFAOYSA-N
Synonym Dodecanamide
PubChem CID 14256
ChEBI CHEBI:34726
IUPAC Name dodecanamide
SMILES CCCCCCCCCCCC(=O)N

Specifications

Melting Point 99°C
Color White-Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 199.34
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Lauramide
EINECSNumber (2)-0824
TSCA Yes