missing translation for 'onlineSavingsMsg'
Learn More

Latanoprost 98.0+%, TCI America™

Catalog No. L026250MG
Change view
Click to view available options
Quantity:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L026250MG 50mg
L026210MG 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L026250MG Supplier TCI America Supplier No. L026250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130209-82-4
Molecular Formula C26H40O5

Specifications

Quantity 50mg
Formula Weight 432.60
Physical Form Clear Liquid at 20°C
Color Yellow
Purity ≥98.0% (HPLC)
Chemical Name or Material Latanoprost
Recommended Storage Freezer