accessibility menu, dialog, popup

UserName

CAS RN 130209-82-4

IUPAC Name: propan-2-yl (5E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate
Synonyms: latanaprost xalatan fixed flow device pharmacia brand of latanoprost latanoprost (product) PhXA34 latanoprost preparation latanoprost (substance)
Molecular Weight (g/mol): 432.6
Molecular Formula: C26H40O5
InChi Key: GGXICVAJURFBLW-RDSJPUOVSA-N
SMILES: CC(C)OC(=O)CCC\C=C\C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1CC[C@@H](O)CCC1=CC=CC=C1
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)