missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Latanoprost

Catalog No. AC466660250
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466660250 25 mg
AC466661000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466660250 Supplier Thermo Scientific Chemicals Supplier No. 466660250

CAS: 130209-82-4 | C26H40O5, C26H40O5 | 432.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130209-82-4
Molecular Formula C26H40O5

Specifications

Quantity 25 mg
Solubility Solubility aq. soln.: insoluble. Other solubilities: soluble in ethanol and acetonitrile
Formula Weight 432.6
InChI Key GGXICVAJURFBLW-RDSJPUOVSA-N
SMILES CC(C)OC(=O)CCC\C=C\C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1CC[C@@H](O)CCC1=CC=CC=C1
IUPAC Name propan-2-yl (5E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate
Molecular Weight (g/mol) 432.60
Chemical Name or Material Latanoprost
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only