missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lasu1/Ureb1 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The ARF tumor suppressor is a critical regulator of p53 stability. In addition to p53, ARF1 binds to other proteins such as MDM2 and ARF-BP1, a large protein containing HECT, UBA and WWE motifs. ARF-BP1 directly binds and ubiquitinates p53; this activity is inhibited by ARF, indicating that ARF-BP1 is a critical mediator of the p53-dependent and p53-independent tumor suppressor functions of ARF. ARF-BP1 can also catalyze the polyubiquitination of Mcl-1, an anti-apoptotic Bcl-2 family member involved in DNA damage-induced apoptosis. Elimination of ARF-BP1 expression by RNA interference stabilized Mcl-1 protein, resulting in an attenuation of apoptosis induced by DNA-damage agents.
Spécifications
Spécifications
| Antigène | Lasu1/Ureb1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | HUWE1 |
| Numéro d’acquisition du gène | Q7Z6Z7 |
| Alias du gène | RP3-339A18.4, ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, UREB1; ARF binding protein 1; ARF-binding protein 1; BJ-HCC-24 tumor antigen; E3 ubiquitin-protein ligase HUWE1; HECT domain protein LASU1; HECT, UBA and WWE domain-containing protein 1; Mcl-1 ubiquitin ligase E3; URE-B1; URE-binding protein 1; homologous to E6AP carboxyl terminus homologous protein 9; large structure of UREB1; upstream regulatory element-binding protein 1 |
| Symboles de gène | HUWE1 |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |