Learn More
GA International Laser & Inkjet Matte Paper Labels 6"" x 0.75"" / 152.4mm x 19.1mm Rectangle White From -29C to +120C (-20F to +248F)
Numéro de catalogue. 5000571223
Encompass
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Numéro de catalogue. 5000571223 Fournisseur GA International Code fournisseur LIP18WH
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.