GA International Laser & Inkjet Matte Paper Labels 4"" x 3.33"" / 101.6mm x 84.6mm Rectangle Purple From -29C to +120C (-20F to +248F)
Numéro de catalogue. 5000571828
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Numéro de catalogue.
5000571828
Fournisseur
GA InternationalCode fournisseur
LIP15PU
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.