Learn More
United Scientific Supplies Laser Diffraction Kit and Accessories
Description
- Explore the mathematics behind diffraction and interference patterns
- Sturdy base securely holds included red and green laser pointers
- Project various patterns against bare walls
- Includes Diffraction Grating (1), Single-Slit Slide (1), Double-Slit Slides (2), Activity Guide
Spécifications
Spécifications
| Product Type | Kit, Laser Diffraction Complete |
| For Use With (Application) | Exploring The Mathematics Behind Diffraction and Interference Patterns |
| For Use With (Equipment) | Laser Diffraction Kit |
| Includes | Diffraction Grating (1), Single-Slit Slide (1), Double-Slit Slides (2), Activity Guide |