Learn More
Advanced Biomatrix LAP Photoinitiator, non-sterile 500 mg
Numéro de catalogue. 502255920
Encompass
LAP is a water soluble photoinitiator for use in the polymerization of hydrogels or bioinks. LAP is photocrosslinked with 405nm blue light and is often preferred over Irgacure 2959 due to improved cytocompatibility.
Numéro de catalogue. 502255920 Fournisseur Advanced Biomatrix Code fournisseur 5269500MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
LAP is a water soluble photoinitiator for use in the polymerization of hydrogels or bioinks. LAP is photocrosslinked with 405nm blue light and is often preferred over Irgacure 2959 due to improved cytocompatibility.