missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LAP (Latency Associated peptide) Monoclonal Antibody (FNLAP), PerCP-eFluor™ 710, eBioscience™
GREENER_CHOICE

Numéro de catalogue. 46982942
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
46982942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 46982942 Fournisseur Invitrogen™ Code fournisseur 46982942
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Description: The FNLAP monoclonal antibody reacts with human latency associated peptide (LAP, pro-TGF beta 1, LAP/TGF beta 1). Many different cells produce TGF beta and it mediates effects on the proliferation, differentiation and function of many cell types. TGF beta is synthesized as a precursor that contains LAP at the N-terminus and mature TGF beta at the C-terminus. Processing and cleavage of the precursor protein between amino acids 278 and 279 results in the formation of LAP dimers and TGF beta dimers that then non-covalently associate with each other to form the small latent TGF beta complex. LAP is secreted and can be found in the extracellular matrix. In addition, LAP can also be expressed on platelets and activated regulatory T cells. It is believed that this surface-expressed LAP is due to the binding of LAP to GARP (LRRC32), which is a transmembrane protein that is also found at high levels on platelets and activated regulatory T cells. Applications Reported: This FNLAP antibody has been reported for use in flow cytometric analysis. Applications Tested: This FNLAP antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits...

TGF beta-1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its prodomain. Activation of TGF-b1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF-beta, an aminoterminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP.

Spécifications

Antigène LAP (Latency Associated peptide)
Applications Flow Cytometry
Classification Monoclonal
Clone FNLAP
Concentration 5 μL/Test
Conjugué PerCP-eFluor 710
Formule PBS with BSA and 0.09% sodium azide; pH 7.2
Gène TGFB1
Numéro d’acquisition du gène P01137
Alias du gène CED; DPD1; LAP; latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGFB; Tgfb1; Tgfb-1; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1; transforming growth factor-beta 1
Symboles de gène TGFB1
Espèces hôtes Mouse
Immunogène E. coli derived hLAP-TGFb, mixed with VB3A9.
Méthode de purification Affinity chromatography
Quantité 100 Tests
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 7040
Espèces cibles Human
Contenu et stockage 4°C, store in dark, DO NOT FREEZE!
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus de résultats Afficher moins de résultats