missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LAP
Description
LAP is a photocrosslinker for hydrogels.
Spécifications
Spécifications
| CAS | 85073-19-4 |
| Quantity | 500 mg |
| Molecular Formula | C16H16LiO3P |
| Formula Weight | 294.21g/mol |
| Product Type | Inhibitor |
For Research Use Only