Learn More
Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate DFS Item
As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development.
Supplier:  Invitrogen™ PV3581
 View more versions of this product
Catalog No. PV3581
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.
Specifications
Specifications
LanthaScreen™ Amine Reactive Tb Chelate, 1mg, 1050.88g/mol Molecular Weight, ≥90% by HPLC Purity, Store at –20°C, Used For Pharma and Biopharma, Target and Lead Identification and Validation
1 mg
LanthaScreen™ TR-FRET labeling reagents are provided lyophilized
≥90% by HPLC
Dry Ice
Drug discovery & development, target & lead identification & validation, TR-FRET assay development
Amine Reactive Tb Chelate
1050.88g/mol
LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day.
LanthaScreen
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.