Learn More
Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate
Description
As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.
Specifications
Specifications
| Description | LanthaScreen™ Amine Reactive Tb Chelate, 10μg, 1050.88g/mol Molecular Weight, ≥90% by HPLC Purity, Store at –20°C, Used For Pharma and Biopharma, Target and Lead Identification and Validation |
| For Use With (Application) | Drug discovery & development, target & lead identification & validation, TR-FRET assay development |
| Quantity | 10 μg |
| Product Type | Amine Reactive Tb Chelate |
| Kit Contents | LanthaScreen™ TR-FRET labeling reagents are provided lyophilized |
| Molecular Weight (g/mol) | 1050.88g/mol |
| Purity | ≥90% by HPLC |
| Content And Storage | LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day. |
| Shipping Condition | Dry Ice |
| Product Line | LanthaScreen |
For Research Use Only. Not for use in diagnostic procedures.