missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Standard, 1000μg/mL in 2-5% HNO3 (Assurance Grade), SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLLA22X

 View more versions of this product

Catalog No. PLLA2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
La1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Lanthanum Standard
7439-91-0 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00011066
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
FZLIPJUXYLNCLC-UHFFFAOYSA-N
2% HNO3
138.91
23926
Plasma
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
0.1,2.0,97.9
La
AA, ICP
Certified Reference Material
[La]
lanthanum
500 mL
CHEBI:33336
Matrix Concentration Cat. No.
2-5% HNO3 1000μg/mL PLLA2-2Y, -2X

Chemical Identifiers

7439-91-0
138.91
FZLIPJUXYLNCLC-UHFFFAOYSA-N
CHEBI:33336
[La]
La
MFCD00011066
23926
lanthanum
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only